Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid ≥99.0%

L(+)-Ascorbic acid ≥99.0%

25 g
Catalog # TCA0537-025G
Supplier:   TCI America
Supplier #:  A0537-25G
L(+)-Ascorbic acid ≥99.0%
Catalog # TCA0537-025G
Supplier:  TCI America
CAS Number:  50-81-7
L(+)-Ascorbic acid ≥99.0%

25 g

CAS Number:  50-81-7
Price
$18.00
Log in for availability
Each (25g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 50-81-7
MDL Number: MFCD00064328
Molecular Formula: C6H8O6
Molecular Weight: 176.12
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 192
Specific rotation [a]20/D: 21 deg (C=10, H2O)

Product Categories: Bioscience, Sugar Acids, Sugars (Others), Regenerative Medicine Research Reagents, Antioxidants, ROS/RNS Detection Reagents, Vitamins
Other Product Names / Synonyms: Vitamin C

Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:192 °C
  • Density:1.694 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Purity/Analysis Method:
    >99.0% (T)

Specifications

  • Size:
    25 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.