$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
GLOVES SURGCL LATX SZ8 PK40PR
PH2D7255
:
Cardinal Health
GLOVES SURGCL LATX SZ8 PK40PR
PH2D7255
:
Cardinal Health
:
2D7255