Orders LinkContactUsLinkComponent
2-Phenoxyethanol
2-Phenoxyethanol
500 ml, Glass bottle, amber, narrow mouth
  JTT319-7
Supplier #:  T319-07
2-Phenoxyethanol
  JTT319-7
 :  122-99-6
2-Phenoxyethanol
2-Phenoxyethanol
500 ml, Glass bottle, amber, narrow mouth
 :  122-99-6
$111.96
Each (500ml) Case of 12 (500ml)
 :  ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
  •  :122-99-6
  •  :C₆H₅OCH₂CH₂OH
  •  :MFCD00002857
  •  :138.17 g/mol
  •  :12,07410
  •  :245 °C (1013 hPa)
  •  :13 °C
  •  :1.1094 g/cm³ (20 °C)
  •  :Management of Change (MOC) category = N
  •  :121 °C (closed cup)
  •  :Ambient
  •  :204-589-7
Documents
  • Identification (by IR):
    Passes Test
  • Pack type:
    Glass bottle, amber, narrow mouth
  • Size:
    500 ml