D-(-)-Mannitol, powder, BAKER ANALYZED® ACS
PE pail, screw top, 12 kg
Supplier: AVANTOR PERFORMANCE MATERIALS US
Supplier #: 2554-07
D-(-)-Mannitol, powder, BAKER ANALYZED® ACS
Catalog # JT2554-7
Supplier: AVANTOR PERFORMANCE MATERIALS US
CAS Number: 69-65-8
MOC: Non-RegulatedCertificates
Jump to:
Product Details & Documents
Powder. Lot analysis on label.
Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:69-65-8
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064287
- Molecular Weight:182.17 g/mol
- Merck Index:13,05769
- Boiling Pt:290…295 °C (4 hPa)
- Melting Pt:164…169 °C
- Density:1.49 g/cm³ (20 °C)
- :Management of Change (MOC) category = N
- Storage Temperature:Ambient
- EINECS:200-711-8
Documents
Certificates
Search by Lot Number2554 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Brand Definition Baker Analyzed
Datasheet
sds_BAKR2554
SDS
Specifications
Specification Test Results
- ACS - Specific Rotation [Å]²⁵ ^D (+):23.3 - 24.3 Degree
- ACS - Insoluble Matter:≤0.01 %
- ACS - Loss on Drying at 105°C:≤0.05 %
- ACS - Residue after Ignition:≤0.01 %
- ACS - Titrable Acid (meq/g):≤0.0008
- ACS - Reducing Sugars:Passes Test
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
Specifications
- Pack type:PE pail, screw top
- Size:12 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



