Chemical Oxygen Demand (COD) Vials
Product Details & Documents
COD vials are used to measure the chemical oxygen demand of influent and effluent of wastewater treatrement facilities. Requires a digestor block and photometer/spectrophotometer.
- Leakproof reagent vials
- Fast safe and easy COD determination
- Works in our COD Photometer or any spectrophotometer accepting a 16 mm round cell
A fresh reagent ampoule blank must be prepared for each series of tests, therefore the number of samples that can be tested with each kit will vary.
Ordering information: COD standards are available in two ranges to run at known concentrations to verify your tests. Single Analyte Meters (SAMs) are available for photometric analysis of the COD vials.
Delivery information: Kits come in a cardboard box and contain everything needed to perform the tests (except distilled water) and instruction book.
Caution: These kits require the use of a Digestor Block along with a CHEMetrics Photometer, a COD Photometer, or a spectrophotometer capable of accepting a 16 mm round cell. All these instruments are sold separately. Unless indicated as mercury-free, the kits contain mercury. Dispose according to local, state or federal laws.
Documents
CHEMetrics Water Quality Solutions for Municipal Water Treatment.pdf CHEMetrics Water Quality Solutions for Pharmaceutical and Biotechnology.pdf
Product Literature
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specifications
- Chemical method:Dichromate reactor digestion
- Description:COD Vial Kit, 25 vials
- Range:0 - 15000 ppm
- Type:COD Vials
- No. of tests:24
Specifications
Accessories
Product Information
- Catalog #DetailsAvailabilityPrice
- COD vial rack
Product Information
- Catalog #RangeTypeAvailabilityPrice
- COD standard, 200 ml
Product Information
- Catalog #AvailabilityPrice
- Photometer for low range COD vials, Range: 0 - 150 ppmPhotometer for low range COD vials, Range: 0 - 150 ppm77674-578Log in for availabilityEach - $773.43
Product Information
- Catalog #AvailabilityPrice
- Photometer for medium to high range COD vials, Range: 0 - 1500 ppm and 0 - 15000 ppmPhotometer for medium to high range COD vials, Range: 0 - 1500 ppm and 0 - 15000 ppm77674-580Log in for availabilityEach - $773.43
Product Information
- Catalog #RangeTypeAvailabilityPrice
- COD standard, 200 ml
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



