$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
CLASS 0 SS WEIGHT SET (1MG-1
AP400002-200
:
Rice Lake Weighing
CLASS 0 SS WEIGHT SET (1MG-1
AP400002-200
:
Rice Lake Weighing
:
45930