$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
CLASS O 5MG WEIGHT KIT
AP400002-194
:
Rice Lake Weighing
CLASS O 5MG WEIGHT KIT
AP400002-194
:
Rice Lake Weighing
:
45882