Ammonia solution 28%
Product Details & Documents
CAS No.: 1336-21-6 Molecular Formula: H5NO Formula Weight: 35.05 Odor: Ammonia-like pH: 12 Merck Reference: 14,494 MDL No.: MFCD00011418
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- Merck Index:14,494
- Boiling Pt:32 - 37.7 °C (1013 hPa)
- Melting Pt:–60…–57.5 °C
- Density:0.90
- Storage Temperature:Ambient
- EINECS:215-647-6
- UN:2672
- ADR:8,III
Documents
Certificates
Search by Lot NumberVWR GHS Compliance Letter (US)
Additional Product Details
VWR® Focus Production Vol 1 Issue 2
Product Literature
Specifications
Specifications
- Pack type:bottle
- Size:500 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



