Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Fumaric acid 99%

Fumaric acid 99%

Bottle, 500 g
Catalog # AAA10976-36
Irritant
Supplier #:  A10976-36
Fumaric acid 99%
Catalog # AAA10976-36
CAS Number:  110-17-8
Fumaric acid 99%

Bottle, 500 g

CAS Number:  110-17-8
Price
$48.51
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

MDL: MFCD00002700 Beilstein Registry No.: 605763 Fieser: 5,319

Purity: 99% CAS No.: 110-17-8 Molecular Formula: HO2CCH=CHCO2H Formula Weight: 116.07 Flash Point: 273°(523°F) Odor: Odorless pH: 2.1 Melting Point: ca 299° subl. Beilstein Reference: 605763 Merck Reference: 14,4287 MDL No.: MFCD00002700

Synonyms:  trans-1,2-Ethylentricarboxylic acid
  • CAS Number:110-17-8
  • Formula:C₄H₄O₄
  • MDL Number:MFCD00002700
  • Molecular Weight:116.07 g/mol
  • Merck Index:14,4287
  • Boiling Pt:356 °C (1013 hPa)
  • Melting Pt:∼ 299 °C (subl.)
  • Density:1.625
  • Flash Pt:273 °C (523 °F)
  • Storage Temperature:Ambient
  • EINECS:203-743-0
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.