Caffeine 99%
Product Details & Documents
Has inhibitory action against lung and colon tumorigenses, and virus induction
Purity: 99% CAS No.: 58-08-2 Molecular Formula: C8H10N4O2 Formula Weight: 194.19 Physical Form: Powder Density: 1.230 Appearance: White Melting Point: 234-236° Solubility: Soluble in pyrrole, THF-H2O mixture, ethyl acetate Beilstein Reference: 17705 Merck Reference: 14,1636 UN No.: 1544 MDL No.: MFCD00005758
Has inhibitory action against lung and colon tumorigenses, and virus induction
- CAS Number:58-08-2
- Formula:C₈H₁₀N₄O₂
- MDL Number:MFCD00005758
- Molecular Weight:194.19 g/mol
- Merck Index:14,1636
- Melting Pt:234…236 °C
- Density:1.230
- Storage Temperature:Ambient
- EINECS:200-362-1
- UN:1544
- ADR:6.1,III
Documents
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specifications
- Pack type:Bottle
- Size:250 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



