Orders LinkContactUsLinkComponent
VWR® 2XYT Medium Broth
VWR® 2XYT Medium Broth
2XYT MEDIUM BROTH, 100 g
Catalog # 97063-440
VWR® 2XYT Medium Broth
Catalog # 97063-440
Supplier:  
CAS Number:  
VWR® 2XYT Medium Broth
VWR® 2XYT Medium Broth
2XYT MEDIUM BROTH, 100 g
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
2XYT BROTH DEHYDR CULTURE MEDIA 500GM Catalog #10053-694
Product 10053-694 is similar to 97063-440 but manufacturer and specifications may vary.
SELECT AGAR 500GM Catalog #90005-196
Product 90005-196 is similar to 97063-440 but manufacturer and specifications may vary.
MEDIUM 2XYT BROTH BIOTECH GRADE 500G Catalog #97063-442
Product 97063-442 is similar to 97063-440 but manufacturer and specifications may vary.
Product Details & Documents
2XYT Medium Broth powder is used to prepare rich broth for the propagation of M13 bacteriophage.
  • Save time using premixed and ready-to-use media
  • Homogenous and high quality powders
  • Light beige, free flowing powder
31g of powder when dissolved per liter of deionized water will yield formulation- 16 grams/L of cesein peptone, 10 grams/L of casamino acids and 5 grams/L of sodium chloride.
Documents
Specifications
Specifications
  • Description:
    2XYT MEDIUM BROTH
  • Size:
    100 g