Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Wright-Giemsa stain solution

Wright-Giemsa stain solution

1 gal.
Catalog # 95057-518
Supplier #:  ES923
Wright-Giemsa stain solution
Catalog # 95057-518
CAS Number:  
Wright-Giemsa stain solution

1 gal.

CAS Number:  
Price
$589.63
Log in for availability
Case of 4 (1/Itm/Bx)
Product Details & Documents

Wright-Giemsa stain is a modified Romanowsky stain used for staining the cellular elements in blood and bone marrow

  • Provides excellent differential staining results
  • Stains RBC inclusion bodies, parasites and other organisms in blood smears
  • Sutiable for automated or manual staining methods
Synonyms:  Wright-Giemsa Solution
  • MDL Number:MFCD00132593
  • Storage Temperature:Ambient

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Size:
    1 gal.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.