Orders LinkContactUsLinkComponent
Capinsert™ Color Coding Inserts, Simport Scientific
Capinsert™ Color Coding Inserts, Simport Scientific
Cap Insert, Lilac
Catalog # 95038-744
Supplier:   Simport Scientific
Supplier #:  T345L
Capinsert™ Color Coding Inserts, Simport Scientific
Catalog # 95038-744
Supplier:  Simport Scientific
Capinsert™ Color Coding Inserts, Simport Scientific
Capinsert™ Color Coding Inserts, Simport Scientific
Cap Insert, Lilac
Price
$33.26
Log in for availability
Case of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Capinserts™ can be used for all color coding needs.
  • Made of polypropylene
  • With write-on frosted area
  • Available in ten colors
The Capinsert™ is used to color code a multitude of Simport® products according to your specific needs. It is inserted on top of the closure and has a write-on frosted area for sample identification.
Specifications
  • Color:
    Lilac
  • Description:
    Cap Insert