Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
DISINF HI-CON 256 NPD 1GL
93001-998
:
Cani
DISINF HI-CON 256 NPD 1GL
93001-998
:
Cani
:
CAN-H256-1
ghs_CAN-H256
VWR GHS Compliance Letter (US)