Orders LinkContactUsLinkComponent
Difco™ Modified mTEC Agar, BD
  90000-796
Difco™ Modified mTEC Agar, BD
  90000-796
 :  
 :  
Difco™ Modified mTEC Agar, BD
 :  
Modified mTEC Agar is a selective culture medium used for the chromogenic detection and enumeration of thermotolerant Escherichia coli in water by the membrane filtration technique.
  • Peptone is a source of nitrogen, amino acids, and carbon
  • Agar is the solidifying agent
Sodium lauryl sulfate and sodium desoxycholate are selective against Gram-positive bacteria. Red to magenta colonies may be presumptively identified as
E. coli.