ELGA Deionization Systems
Deionizer replacement cartridge, for Vision 125, filter
Catalog # 89204-278
Supplier: ELGA LabWater
Supplier #: VC125
Jump to:
Product Details & Documents
ELGA’s range of wall mounted deionization systems produce from 30 to 90 liters per hour of high purity water for a wide range of laboratory and general manufacturing applications.
- Cost-effective
- Easy monitoring
- Built-in meters
- Water Quality: Type II
The deionizers are cost-effective stand-alone systems that provide low to medium volumes of primary or deionized grade water. Water quality is easy to monitor, with built in meters or colour change indicators.
Documents
Specifications
- Description:Deionizer replacement cartridge, for Vision 125, filter