ELGA Deionization Systems
Vision 250 deionizer, 92×110×660 mm (3.6×4.3×26"), 60 LPH, 4.2 kg (9.3 lbs.)
89203-952
ELGA’s range of wall mounted deionization systems produce from 30 to 90 liters per hour of high purity water for a wide range of laboratory and general manufacturing applications.
- Cost-effective
- Easy monitoring
- Built-in meters
- Water Quality: Type II
The deionizers are cost-effective stand-alone systems that provide low to medium volumes of primary or deionized grade water. Water quality is easy to monitor, with built in meters or colour change indicators.
- Flow rate:60 LPH
- Description:Vision 250 deionizer
- W×D×H:92×110×660 mm (3.6×4.3×26")
- Weight:4.2 kg (9.3 lbs.)



