Orders LinkContactUsLinkComponent
SlideFolder™ Microscope Slide Holders, Simport Scientific
295×192×11 mm (11 11/16×7 9/16×7/16”), Yellow
  89039-694
 :  M750-20Y
SlideFolder™ Microscope Slide Holders, Simport Scientific
  89039-694
SlideFolder™ Microscope Slide Holders, Simport Scientific
295×192×11 mm (11 11/16×7 9/16×7/16”), Yellow
$224.02
Case of 10
These two-part folders hold up to twenty standard microscope slides
  • All units are stackable
  • Not autoclavable
The high-impact polystyrene base holds slides horizontally with numbered spaces for easy identification. The transparent polystyrene door covers the slides and can be swung behind the folder for space-saving purposes.
Temperature range: –80 to 80°C (–112 to 176°F)
  • Color:
    Yellow
  • Dimensions:
    295×192×11 mm (11 11/16×7 9/16×7/16”)