Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Dropper® Spinal Fluid Control, Quantimetrix

Dropper® Spinal Fluid Control, Quantimetrix

Dropper® Spinal Fluid Control, Level 1, 3× 3 ml
Catalog # 82024-162
Supplier:   Quantimetrix
Supplier #:  1451-31
Dropper® Spinal Fluid Control, Quantimetrix
Catalog # 82024-162
Supplier:  Quantimetrix
CAS Number:  
Dropper® Spinal Fluid Control, Quantimetrix
Dropper® Spinal Fluid Control, Quantimetrix

Dropper® Spinal Fluid Control, Level 1, 3× 3 ml

CAS Number:  
Price
$192.06
Log in for availability
Box of 3 (3ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Dropper Spinal Fluid Control monitors the performance of spinal fluid chemistry assay methods. Drop the control directly into the instrument sample cup with an easy-to-use, dropper bottle for simple dispensing and maximum use. Dropper Spinal Fluid Control is designed for use with most major chemistry analyzers and nephelometers including Beckman Coulter Synchron®, Siemens Dimension®, Beckman Coulter AU® series, Ortho Vitros®, Roche Cobas®, Abbott Arcitect and Pyrogallol Red microprotein methods.

  • Easy-to-use dropper bottles
  • Liquid, human protein matrix
  • 2-year refrigerated shelf life from date of manufacture
  • 90-day open vial stability when stored at 2 to 8°C

Ordering information: Refer to the package insert of currently available lots for specific analyte, stability claims and instrument compatibility.

Documents

Specifications

Specifications

  • Description:
    Dropper® Spinal Fluid Control
  • Volume:
    3× 3 ml
  • Includes:
    Level 1

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.