Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER HIGH RESOLUTION
Catalog # 82009-939
Supplier:
Apacor
Expand image
BUFFER HIGH RESOLUTION
Catalog # 82009-939
Supplier:
Apacor
Supplier Number:
51104