Poly(caprolactone), soluble
Product 700012-648 is similar to 700012-646 but manufacturer and specifications may vary.
Product Details & Documents
Poly(caprolactone), Flakes is a biodegradable polymer often used as an additive for resins to improve their processing characteristics and their end use properties (e.g., impact resistance).
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
- CAS Number:24980-41-4
- Formula:(C₆H₁₀O₂)n
- MDL Number:MFCD00084404
- Molecular Weight:114.1400 g/mol
- Density:1.146
- Storage Temperature:Ambient
Documents
Specifications
Specifications
- Pack type:Poly bottle
- Size:250 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



