Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
γ-Cyclodextrine ≥98% suitable for use as excipient

γ-Cyclodextrine ≥98% suitable for use as excipient

Glass, amber, 100 mg
Catalog # 700011-306
Supplier #:  C1623-100MG
γ-Cyclodextrine ≥98% suitable for use as excipient
Catalog # 700011-306
CAS Number:  17465-86-0
γ-Cyclodextrine ≥98% suitable for use as excipient

Glass, amber, 100 mg

CAS Number:  17465-86-0
Price
$171.88
Log in for availability
Each (100mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

gamma-Cyclodextrin, also known as Cyclodextrins, are used in the food, pharmaceutical, and chemical industries because they enhance the solubility and bioavailability of such compounds. This is of high interest for pharmaceutical as well as dietary supplement applications in which hydrophobic compounds shall be delivered. Alpha-, beta-, and gamma-cyclodextrin are all generally recognized as safe by the FDA.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonyms:  gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
  • CAS Number:17465-86-0
  • Formula:C₄₈H₈₀O₄₀
  • MDL Number:MFCD00009595
  • Molecular Weight:1297.1248 g/mol
  • Storage Temperature:Ambient
  • EINECS:241-482-4
Specifications

Specification Test Results

  • Assay (%):
    98

Specifications

  • Pack type:
    Glass, amber
  • Size:
    100 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.