Orders LinkContactUsLinkComponent
Round Bottom Flasks
150 ml, 19/26, Round
Catalog # 47800-124
Supplier:   Heidolph NA, LLC
 :  015890610
Round Bottom Flasks
Catalog # 47800-124
Supplier:  Heidolph NA, LLC
Round Bottom Flasks
150 ml, 19/26, Round
Price
$430.48
Log in for availability
Pack of 5
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Specifications
  • [ST] joint no.:
    19/26
  • Capacity:
    150 ml
  • Bottom style:
    Round