Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Corning® Trypsin

Corning® Trypsin

0.05% Trypsin 0.53 mM EDTA, 100 ml
Catalog # 45001-082
Supplier:   Corning
Supplier #:  25-051-CI
Corning® Trypsin
Catalog # 45001-082
Supplier:  Corning
CAS Number:  
Corning® Trypsin
Corning® Trypsin

0.05% Trypsin 0.53 mM EDTA, 100 ml

CAS Number:  
Price
$75.80
Log in for availability
Pack of 6
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging. Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances the efficacy of trypsin.

  • Storage: -25°C to -15°C

Trypsin has a pH specification range from 7.2 to 8. At this range, phenol red inclusion gives the solution a pink color. During shipping, the trypsin might change color from pink to yellow due to exposure to carbon dioxide released from the dry ice in the shipping container. In our experience, this discoloration does not affect the functionality of the trypsin. By adjusting the pH to 7.2 – 8 with NaOH the trypsin activity could be restored.

Specifications

Specifications

  • Description:
    0.05% Trypsin 0.53 mM EDTA
  • Size:
    100 ml

Specifications