Corning® Lymphocyte Separation Medium, Corning
Lymphocyte Separation Medium, 100 ml
Supplier: Corning
Supplier #: 25-072-CI
Jump to:
Product Details & Documents
Lymphocyte Separation Medium (LSM) was originally designed for the in vitro isolation of lymphocytes from diluted whole blood
It is a sterile, iso-osmotic polysucrose and diatrizoate solution with low viscosity and a density of 1.077-1.080 g/mL at 20ºC.
Specifications
- Volume:100 ml
- Description:Lymphocyte Separation Medium
- Storage Conditions:Ambient
- Cat. No.:45000-726
- Shipping Conditions:Ambient
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



