Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
MEM, Corning® 50 L MEM (Minimum Essential Medium) alpha medium, powder [+] Earles salts, L-glutamine [-] ribonucleosides, deoxyribonucleosides, sodium bicarbonate, Powder, 50 L
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
Catalog # 45000-504
CAS Number:
Jump to:
Discontinued
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells.
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Liquid media products are formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts.
Certifications: Triple 0.1 µm sterilized.
Documents
Certificates
Search by Lot Number3D Cell Culture Quick Reference Ordering Guide Corning 3D Cell Culture Quick Reference Ordering Guide Corning 3D Cell Culture.pdf Corning® Shopping list for returning to the bench
Product Literature
Cell Culture Media, Molecular Buffers and Reagents
Additional Product Details
Specifications
Specifications
- Media Format:Powder
- Description:MEM, Corning® 50 L MEM (Minimum Essential Medium) alpha medium, powder [+] Earles salts, L-glutamine [-] ribonucleosides, deoxyribonucleosides, sodium bicarbonate
- Size:50 L
- Cat. No.:45000-504
- Storage Temperature:2...8 °C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



