Orders LinkContactUsLinkComponent
Sampling Tubes and Systems, Drager
Diffusion sampler ORSA 5
  34330-184
 :   Draeger
 :  6728891
Sampling Tubes and Systems, Drager
  34330-184
 :  Draeger
Sampling Tubes and Systems, Drager
Sampling Tubes and Systems, Drager
Diffusion sampler ORSA 5
$240.09
Each
Using the Drager sampling system, hazardous substances in the air are collected using a suitable medium via adsorption or chemisorption.
In active sampling, the air to be evaluated is drawn through a sampling tube such as Activated Charcoal and Silica Gel tube types with a pump and passive sampling is performed with diffusion samplers like the Dräger ORSA or the Dräger Nitrous Oxide diffusion samplers.
  • Description:
    Diffusion sampler ORSA 5