Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ECO LINE FLUX REMOVER 5-GAL
Catalog # 14220-324
Supplier:
ITW Chemtronics
SDS
Expand image
ECO LINE FLUX REMOVER 5-GAL
Catalog # 14220-324
Supplier:
ITW Chemtronics
Supplier Number:
1621-5G
SDS
Related Information
Documentation
SDS
ghs_1621
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation