Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ISOPROPYL ALCOHOL 54 GAL 54GL
Catalog # 14220-318
Supplier:
ITW Chemtronics
SDS
Expand image
ISOPROPYL ALCOHOL 54 GAL 54GL
Catalog # 14220-318
Supplier:
ITW Chemtronics
Supplier Number:
1610-54G
SDS
Related Information
Documentation
SDS
ghs_1610
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation