Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CAL GAS CO 250PPM 103L IN AIR
Catalog # 13453-514
Supplier:
Draeger
SDS
Expand image
CAL GAS CO 250PPM 103L IN AIR
Catalog # 13453-514
Supplier:
Draeger
Supplier Number:
4502154
SDS
Related Information
Documentation
SDS
ghs_4502154.pdf
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation