Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CAL GAS HCN 10PPM 58L IN N2
Catalog # 13453-508
Supplier:
Draeger
SDS
Expand image
CAL GAS HCN 10PPM 58L IN N2
Catalog # 13453-508
Supplier:
Draeger
Supplier Number:
4594962
SDS
Related Information
Documentation
SDS
ghs_4594962.pdf
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation