Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
qScript™ cDNA Synthesis Kits, QuantaBio

qScript™ cDNA Synthesis Kits, QuantaBio

qScript™ cDNA Synthesis Kits, 25
Catalog # 101414-096
Supplier:   Quantabio
Supplier #:  95047-025
qScript™ cDNA Synthesis Kits, QuantaBio
Catalog # 101414-096
Supplier:  Quantabio
CAS Number:  
qScript™ cDNA Synthesis Kits, QuantaBio
qScript™ cDNA Synthesis Kits, QuantaBio

qScript™ cDNA Synthesis Kits, 25

CAS Number:  
Price
$139.96
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The qScript cDNA Synthesis Kit is a sensitive and easy-to-use solution for RNA quantification using two step RT-qPCR.

  • Economical 2-component kit ideally suited for high throughput expression studies
  • Optimized, double pre-primed master mix component ensures equal representation of 5′ and 3′ RNA sequences
  • Broad linear dynamic detection range with limiting (10pg) or plentiful (1ug) samples of total RNA

The qScript™ reaction mix provides all the necessary components for cDNA synthesis except the enzyme and RNA template. The optimized blend of random and oligo(dT) primers provides robust, consistent, and unbiased first-strand synthesis over a broad range of RNA template concentrations. qScript™ reverse transcriptase is a mixture of an engineered MMLV-RT and a ribonuclease inhibitor protein. The simplified reaction procedure is ideally suited for high throughput expression studies using RT-PCR. The resulting cDNA product is directly compatible with current real-time PCR methods or conventional end-point RT-PCR where the targets are ≤1kb in length.

Specifications

Specifications

  • Description:
    qScript™ cDNA Synthesis Kits
  • No. of reactions:
    25

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.