Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Galactose Solution, Teknova

Galactose Solution, Teknova

Galactose Solution, 20%, 250 mL
Catalog # 100217-100
Supplier:   Teknova
Supplier #:  G0505
Galactose Solution, Teknova
Catalog # 100217-100
Supplier:  Teknova
CAS Number:  
Galactose Solution, Teknova

Galactose Solution, 20%, 250 mL

CAS Number:  
Price
$107.10
Log in for availability
Each (250ml)
Minimum order quantity is 4
Product Details & Documents

For used in growth media and stored at room temperature.

Specifications

Specifications

  • Description:
    Galactose Solution, 20%
  • Size:
    250 mL
  • Cat. No.:
    100217-100
  • Sterility:
    Sterile

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.