Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Galactose Solution

Galactose Solution, 20%, 500 mL
Catalog # 100216-666
Galactose Solution
Catalog # 100216-666
Supplier:  
CAS Number:  
Galactose Solution

Galactose Solution, 20%, 500 mL

CAS Number:  
Discontinued
Product Details & Documents

For used in growth media and stored at room temperature.

Specifications

Specifications

  • Description:
    Galactose Solution, 20%
  • Size:
    500 mL
  • Cat. No.:
    100216-666
  • Sterility:
    Sterile

Specifications