Acetone USP Test Solution (TS), buffered
500 ml
Supplier: Ricca Chemical
Supplier #: R0230000-500A
Acetone USP Test Solution (TS), buffered
Catalog # RC230-16
Supplier: Ricca Chemical
CAS Number: 67-64-1
Jump to:
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- UN:1090
- ADR:3,II
Documents
Certificates
Search by Lot Numberghs_R0230000
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specification Test Results
- Appearance:Clear to slight haze, colorless, Acetone odor
- pH at 25 °C (traceable to NIST SRM 186 & 191):5.9 ± 0.2 pH at 25.0 °C
Specifications
- Size:500 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



