$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
GLOVE TRIFLEX VYL PF ST EX SM
PH2D7011PF
:
Cardinal Health
GLOVE TRIFLEX VYL PF ST EX SM
PH2D7011PF
:
Cardinal Health
:
2D7011PF