Orders LinkContactUsLinkComponent
Comparators for CHEMets® Test Kits
Comparators for CHEMets® Test Kits
Nitrite Flat Comparator, 0 - 3000 ppm, 0, 400, 600, 800, 1100, 1500, 1900, 2400, 3000 ppm
Catalog # CXC-7004C
Comparators for CHEMets® Test Kits
Catalog # CXC-7004C
Supplier:  
CAS Number:  
Comparators for CHEMets® Test Kits
Comparators for CHEMets® Test Kits
Nitrite Flat Comparator, 0 - 3000 ppm, 0, 400, 600, 800, 1100, 1500, 1900, 2400, 3000 ppm
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Caution: Store in the dark at room temperature.
Documents
Specifications
Specifications
  • Description:
    Nitrite Flat Comparator
  • Increments:
    0, 400, 600, 800, 1100, 1500, 1900, 2400, 3000 ppm
  • Range:
    0 - 3000 ppm