Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Corning® FluoroBlok™ Multiwell Insert Systems, Corning

Corning® FluoroBlok™ Multiwell Insert Systems, Corning

96, 3.0 µm
Catalog # BD351161
Supplier:   Corning
Supplier #:  351161
Corning® FluoroBlok™ Multiwell Insert Systems, Corning
Catalog # BD351161
Supplier:  Corning
Corning® FluoroBlok™ Multiwell Insert Systems, Corning
Corning® FluoroBlok™ Multiwell Insert Systems, Corning

96, 3.0 µm

Price
$496.00
Log in for availability
Case of 1
Minimum order quantity is 1
Product Details & Documents

Corning® FluoroBlok™ HTS 96-multiwell insert system is a cell culture assay platform designed for automation. The one-piece insert housing and fluorescence-blocking microporous membrane (available in 3.0 and 8.0 μm pore sizes) enables increased efficiency, productivity and throughput in the drug discovery process. The receiver plate design minimizes crosstalk between wells.

  • Standard technology platform allows multiple protocols
  • Real time detection without dismantling or destroying the insert
  • Eliminates need for manual cell scraping and counting
  • Block >99% of the excitation and emission wavelengths of fluorophores commonly used to label cells

The wide blocking range (490 to 700 nm) of the Corning® FluoroBlok™ membrane allows the flexibility to choose from a variety of fluorophores for screening compounds in cell-based assays such as compound permeability, cell invasion and migration, and monolayer permeability. Each insert has an automation compatible sampling port for sampling above and below the membrane with standard pipet tips or automated fluid handling equipment.

The 96-well feeder trays with lids and the 96 square-well flat bottom assay plates feature a nontreated, hydrophobic polystyrene surface. Both are sterile and pyrogen-free.

Systems are designed to increase efficiency, productivity, and throughput. The black housing of the 96-multiwell insert minimizes crosstalk between the wells and virtually eliminates autofluorescence. The multiwell insert plate format is compatible with standard fluorescence plate readers, robots, and fluid handling instruments.

Specifications
  • No. of wells:
    96
  • Pore size:
    3.0 µm
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Additional Feeder Tray with Lid for 24-Well Systems
    Additional Feeder Tray with Lid for 24-Well Systems
    BD351186
    Log in for availability
    Case of 5 - $144.00

Product Information

  • Catalog #
    Availability
    Price
  • Feeder Tray and Lid for 96-Well Systems
    Feeder Tray and Lid for 96-Well Systems
    BD-353924
    Log in for availability
    Case of 5 - $175.00

Product Information

  • Catalog #
    Availability
    Price
  • Square-Well, Flat-Bottom Assay Plates for 96-Well Systems
    Square-Well, Flat-Bottom Assay Plates for 96-Well Systems
    BD353928
    Log in for availability
    Case of 5 - $188.00