BD Eclipse™ Needles and Syringes, BD
Product Details & Documents
BD Eclipse™ Needles are designed with pivoting needle technology for single-handed activation
For use with BD Luer-Lok™ Syringes.
No hard surface is required with this needle-based, safety-engineered injection device.
Documents
Specifications
- Volume:10 ml
- Description:BD Luer-Lok Syringe with Detachable Needle
- Needle length:11/2"
- Gauge:22