Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Behenic acid tech. 85%, Technical Grade

Behenic acid tech. 85%, Technical Grade

bottle, 50 g
Catalog # AAAA12850-18
Behenic acid tech. 85%, Technical Grade
Catalog # AAAA12850-18
Supplier:  
CAS Number:  112-85-6
Behenic acid tech. 85%, Technical Grade

bottle, 50 g

CAS Number:  112-85-6
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BEHENIC ACID 25GM Catalog #TCB1248-025G

Product TCB1248-025G is similar to AAAA12850-18 but manufacturer and specifications may vary.

BEHENIC ACID Catalog #TCB1248-5G

Product TCB1248-5G is similar to AAAA12850-18 but manufacturer and specifications may vary.

BEHENIC ACID 25GM Catalog #TCD0963-025G

Product TCD0963-025G is similar to AAAA12850-18 but manufacturer and specifications may vary.

See more alternatives
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 85% CAS No.: 112-85-6 Molecular Formula: CH3(CH2)20CO2H Formula Weight: 340.60 Melting Point: 74-78° Boiling Point: 306°/60mm Merck Reference: 14,1023 MDL No.: MFCD00002807

Synonyms:  Docosanoic acid
  • CAS Number:112-85-6
  • Formula:C₂₂H₄₄O₂
  • MDL Number:MFCD00002807
  • Molecular Weight:340.59 g/mol
  • Merck Index:14,1023
  • Boiling Pt:306 °C/60 mm
  • Melting Pt:74…78 °C
  • Density:0.8221 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-010-8
Specifications

Specifications

  • Pack type:
    bottle
  • Size:
    50 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.