Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Diethyl ether ≥98% stabilised ACS

Diethyl ether ≥98% stabilised ACS

Bottle, 6×500 g
Catalog # AA33224-62
Flammable
Irritant
Supplier #:  033224-62
Diethyl ether ≥98% stabilised ACS
Catalog # AA33224-62
CAS Number:  60-29-7
Diethyl ether ≥98% stabilised ACS
Diethyl ether ≥98% stabilised ACS

Bottle, 6×500 g

CAS Number:  60-29-7
Price
$244.12
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Widely used as a solvent. Dissolves P, S, Br₂ and I₂

Purity: 98% min CAS No.: 60-29-7 Molecular Formula: C4H10O Formula Weight: 74.12 Flash Point: -45°(-49°F) Physical Form: Liquid Density: 0.713 Refractive Index: 1.3525 Appearance: Colorless Odor: Sweet Melting Point: -116° Boiling Point: 34.6° Solubility: Soluble in H2SO4. Sparingly soluble in water. Miscible in most organic solvents Merck Reference: 14,3806 UN No.: 1155 MDL No.: MFCD00011646

Widely used as a solvent. Dissolves P, S, Br2 and I2

Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Merck Index:14,3806
  • Boiling Pt:34.6 °C
  • Melting Pt:–116 °C
  • Density:0.713
  • Flash Pt:–45 °C (–49 °F)
  • UN:1155
  • ADR:3,I
  •  :Stabilised with BHT (2,6-Di-tert-butyl-p-cresol, 2,6-Di-tert-butyl-4-methylphenol, Ionol) and ethanol (0.001%:3%)
  • Storage Temperature:Ambient
  • EINECS:200-467-2
  • REACH:01-2119535785-29
Specifications

Specification Test Results

  • Color (APHA):
    10
  • Peroxide (as H2O2):
    1ppm ppm
  • Evaporation residue:
    0.001%
  • Titratable acid:
    0.0002meq/g
  • Carbonyl (as HCHO):
    0.001%

Specifications

  • Pack type:
    Bottle
  • Size:
    6×500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.