VWR® Blocking Buffers
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Most routinely used blocking buffers in a variety of formats for convenience.
- Liquid or pre-measured powder Ready-Packs™ available for easy use
- Optimize assay performance using high quality blocking buffers
Each pack of powdered blocking buffer makes one liter of 1X solution.
Documents
Specifications
Specifications
- Description:Fish Gelatin Blocking Buffer, 10%
- Size:500 mL