Hardness 1 Buffer Solution, Hach
Hardness 1 Buffer Solution, 500 ml, EDTA, Bottle
Catalog # 97026-664
Product available on GSA Advantage®
Hardness 1 Buffer Solution, Hach
Catalog # 97026-664
Product available on GSA Advantage®
CAS Number:
Jump to:
Discontinued
Product Details & Documents
Buffer for hardness and chelant determination by titration, pH 10.1 ± 0.1 at 20°C
Hach methods 8213 and 8226. Also used in Hach Phenols method 8047.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Chemical method:EDTA
- Pack type:Bottle
- Description:Hardness 1 Buffer Solution
- Size:500 ml
- Cat. No.:97026-664
- Method:8213, 8226 - EDTA Titration



