Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Wright-Giemsa stain solution

Wright-Giemsa stain solution

32 oz.
Catalog # 95057-514
Supplier #:  ES921
Wright-Giemsa stain solution
Catalog # 95057-514
CAS Number:  
Wright-Giemsa stain solution

32 oz.

CAS Number:  
Price
$77.65
Log in for availability
Each (32oz)
Product Details & Documents

Wright-Giemsa stain is a modified Romanowsky stain used for staining the cellular elements in blood and bone marrow

  • Provides excellent differential staining results
  • Stains RBC inclusion bodies, parasites and other organisms in blood smears
  • Sutiable for automated or manual staining methods
Synonyms:  Wright-Giemsa Solution
  • MDL Number:MFCD00132593
  • Storage Temperature:Ambient

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Size:
    32 oz.

Specifications