Capinsert™ Color Coding Inserts, Simport Scientific
Product Details & Documents
Capinserts™ can be used for all color coding needs.
- Made of polypropylene
- With write-on frosted area
- Available in ten colors
The Capinsert™ is used to color code a multitude of Simport® products according to your specific needs. It is inserted on top of the closure and has a write-on frosted area for sample identification.
Specifications
- Color:Violet
- Description:Cap Insert