Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
BBL™ MacConkey II Agar, BD

BBL™ MacConkey II Agar, BD

MacConkey II agar, 500 g
Catalog # 90000-594
Supplier:   BD
Supplier #:  212306
BBL™ MacConkey II Agar, BD
Catalog # 90000-594
Supplier:  BD
CAS Number:  
BBL™ MacConkey II Agar, BD
BBL™ MacConkey II Agar, BD

MacConkey II agar, 500 g

CAS Number:  
Price
$226.00
Log in for availability
Each (500g)
Product Details & Documents

MacConkey II Agar is a selective and differential medium for the detection of coliform organisms and enteric pathogens.

  • Low concentration of bile salts
  • Crystal violet inhibits growth of gram-positive bacteria
  • Lactose and neutral red indicator aid in differentiation of enteric microorganisms

MacConkey II Agar is recommended for use with clinical specimens likely to contain mixed microbial flora, such as urine, because it allows a preliminary gropuing of enteric and other gram-negative bacteria.

Specifications

Specifications

  • Description:
    MacConkey II agar
  • Size:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.