Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
MacConkey Agar, BD

MacConkey Agar, BD

MacConkey Agar, 500 g
Catalog # 90000-168
Supplier:   BD
Supplier #:  211387
MacConkey Agar, BD
Catalog # 90000-168
Supplier:  BD
CAS Number:  
MacConkey Agar, BD
MacConkey Agar, BD

MacConkey Agar, 500 g

CAS Number:  
Price
$197.00
Log in for availability
Each (500g)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

MacConkey agar is a selective and differential culture medium for bacteria designed to selectively isolate Gram-negative and enteric bacilli and differentiate them based on lactose fermentation.

  • Isolates Gram-negative bacilli

Enteric bacteria that have the ability to ferment lactose can be detected using the carbohydrate lactose, and the pH indicator neutral red.

Specifications

Specifications

  • Description:
    MacConkey Agar
  • Size:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.