$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
REACTION BLOCK LIQUID 16
83007-280
:
Heidolph NA, LLC
REACTION BLOCK LIQUID 16
83007-280
:
Heidolph NA, LLC
:
036190190