Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Corning® Tris Base Buffer, Powder, Corning

Tris Base Buffer, Powder, 1 kg
  62111-158
 :   Corning
 :  61-233-RR
Corning® Tris Base Buffer, Powder, Corning
  62111-158
 :  Corning
 :  
Corning® Tris Base Buffer, Powder, Corning
Corning® Tris Base Buffer, Powder, Corning

Tris Base Buffer, Powder, 1 kg

 :  
 
$677.00
   
Each (1kg)
 
   
 

Tris is commonly used in buffer solutions in biochemical and molecular biological research

This primary amine has an effective pH range of 7.0 - 9.2 and is reported to inhibit a number of different enzymes, limiting its usefulness in protein studies to some extent. This tris base buffer is provided in powder form.

 

 

 

  • Description:
    Tris Base Buffer, Powder
  • Size:
    1 kg
  • Cat. No.:
    62111-158
  • Shipping Temperature:
    Ambient
  • Storage Temperature:
    Ambient

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.