Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Open Handle Probes, Seedburo

Open Handle Probes, Seedburo

Open Handle Probe, Brass, 10 openings, 160 cm (63")
Catalog # 56724-039
Supplier:   Seedburo Equipment
Supplier #:  22-OH
Open Handle Probes, Seedburo
Catalog # 56724-039
Supplier:  Seedburo Equipment
Open Handle Probes, Seedburo
Open Handle Probes, Seedburo

Open Handle Probe, Brass, 10 openings, 160 cm (63")

Price
$1,590.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Open-ended probes without partitions are more convenient to empty and are widely used by the grain elevator operators for obtaining samples from truck loads.

  • Extra heavy gauge tubing for durable, time-tested sampling
  • Held upright and sample is dumped from the handle
Specifications
  • Description:
    Open Handle Probe, Brass, 10 openings
  • Length:
    160 cm (63")